VWR® 950 Benchtop Conductivity Meter
Supplier: VWR International
77618-988 77618-990 77618-992 77618-994
77618-988EA 1366.78 USD
77618-988 77618-990 77618-992 77618-994
VWR® 950 Benchtop Conductivity Meter
Electrochemistry Meters Conductivity Meters
The VWR-EC950 meter provide accurate and reliable measurements of conductivity, resistivity, TDS, salinity and temperature within one versatile meter.  
  • High resolution color touch screen (7.0")
  • Five point calibration with standard recognition
  • IP 54 rating
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Ability to save 1000 data sets for each parameter (GLP-compliant)
  • Settable parameters, including cell constant, temperature compensation coefficient, and TDS factor
  • Various temperature types (none, linear, and pure water)
The easy to use VWR-EC950 benchtop conductivity meter provides an array of measurement parameters to allow for use in a variety of applications. The intelligent operation system provides features including user managment, calibration management, electrode management, method management, and many other advanced capabilities.
Connect the meter easily to the VWR dot matrix printer (10799-132) to print measurement reports.
Información de suministro:
Three-year meter warranty. All meters include: In use cover, touch screen pen, and electrode stand. For application specific recommendations, please refer 'VWR® electrode selection guide' located in the related information section.