Rat alpha 2 Macroglobulin ELISA Kits
Supplier: Abcam
AB320793-1X384TESTS AB320793-1X96TESTS AB320793-10X96TESTS
ABCA_AB320793-1X38EA 2388 USD
ABCA_AB320793-1X38 ABCA_AB320793-1X96 ABCA_AB320793-10X9
Rat alpha 2 Macroglobulin ELISA Kits
Assays ELISAs
Rat alpha 2 Macroglobulin ELISA Kit is a single-wash 90-min Simplestep used to quantify Rat alpha 2 Macroglobulin with a sensitivity of 14 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput .