Human IRAK-1 ELISA Kit
Supplier: Abcam
AB219630-1X384TESTS AB219630-1X96TESTS AB219630-10X96TESTS
ABCA_AB219630-1X38EA 2388 USD
ABCA_AB219630-1X38 ABCA_AB219630-1X96 ABCA_AB219630-10X9
Human IRAK-1 ELISA Kit
Assays ELISAs
Human IRAK-1 ELISA kit is a single-wash 90-min Simplestep used to quantify Human IRAK-1 with a sensitivity of 22 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.