Immunoassay Blocking (BSA Free)
Supplier: Abcam
AB171535-50ML AB171535-1000ML
ABCA_AB171535-50MLEA 107 USD
ABCA_AB171535-50ML ABCA_AB171535-1000
Immunoassay Blocking (BSA Free)
Buffers Electrophoresis Buffers Blocking Buffers
Immunoassay Blocking (BSA Free) (ab171535) maintains the activity of antibodies and other biomolecules adsorbed or immobilized to solid substrates.