Mouse IL-10 ELISA Set (without plates)
Supplier: Abcam
AB47599-10X96TESTS AB47599-5X96TESTS AB47599-15X96TESTS
ABCA_AB47599-10X96EA 1228 USD
ABCA_AB47599-10X96 ABCA_AB47599-5X96T ABCA_AB47599-15X96
Mouse IL-10 ELISA Set (without plates)
Assays ELISAs
Mouse IL-10 ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IL-10 ELISA Set (without plates) in Mouse in Plasma, Cell culture supernatant, Serum samples.