Human IP-10 ELISA Kit
Supplier: Abcam
AB289906-1X384TESTS AB289906-1X96TESTS
ABCA_AB289906-1X38EA 2388 USD
ABCA_AB289906-1X38 ABCA_AB289906-1X96
Human IP-10 ELISA Kit
Assays ELISAs
Human IP-10 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human IP-10 with a sensitivity of 1.039 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.