Human IgG1 ELISA Kit
Supplier: Abcam
AB317543-10X96TESTS AB317543-1X384TESTS AB317543-1X96TESTS
ABCA_AB317543-10X9EA 7858 USD
ABCA_AB317543-10X9 ABCA_AB317543-1X38 ABCA_AB317543-1X96
Human IgG1 ELISA Kit
Assays ELISAs
Human IgG1 ELISA kit is a single-wash 90-min Simplestep used to quantify Human IgG1 with a sensitivity of 6.237 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.