Human Apolipoprotein CI/Apo-CI ELISA Kit- Extracellular
Supplier: Abcam
AB320791-1X96TESTS AB320791-10X96TESTS AB320791-1X384TESTS
ABCA_AB320791-1X96EA 877 USD
ABCA_AB320791-1X96 ABCA_AB320791-10X9 ABCA_AB320791-1X38
Human Apolipoprotein CI/Apo-CI ELISA Kit- Extracellular
Assays ELISAs
Human Apolipoprotein CI/Apo-CI ELISA Kit- Extracellular is a single-wash 90-min Simplestep used to quantify Human Apolipoprotein CI/Apo-CI with a sensitivity of 14.44 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.