Gentamicin ELISA Kit - Intracellular
Supplier: Abcam
AB322085-1X96TESTS AB322085-10X96TESTS
ABCA_AB322085-1X96EA 1009 USD
ABCA_AB322085-1X96 ABCA_AB322085-10X9
Gentamicin ELISA Kit - Intracellular
Assays ELISAs
Gentamicin ELISA Kit - Intracellular is a single-wash 90-min Simplestep used to quantify Gentamicin with a sensitivity of 53.19 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Competitive ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders.