Human IL-1a ELISA Kit
Supplier: Abcam
AB178008-10X96TESTS AB178008-1X96TESTS
ABCA_AB178008-10X9EA 9062 USD
ABCA_AB178008-10X9 ABCA_AB178008-1X96
Human IL-1a ELISA Kit
Assays ELISAs
Human IL-1a ELISA Kit is a single-wash 90-min Simplestep used to quantify Human IL-1a with a sensitivity of 0.82 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders.