Mouse Thrombopoietin ELISA Kit
Supplier: Abcam
AB317546-10X96TESTS AB317546-1X384TESTS AB317546-1X96TESTS
ABCA_AB317546-10X9EA 7858 USD
ABCA_AB317546-10X9 ABCA_AB317546-1X38 ABCA_AB317546-1X96
Mouse Thrombopoietin ELISA Kit
Assays ELISAs
Mouse Thrombopoietin ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Thrombopoietin with a sensitivity of 18.8 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair