Argipressin
Supplier: ALADDIN SCIENTIFIC
A302232-25MG A302232-5MG
ALA302232-25MGEA 207.9 USD
ALA302232-25MG ALA302232-5MG
Argipressin
Proteins and Peptides
Argpressin Acetate is a neurohypophysial hormone found in most mammals. It acts as a neurotransmitter at synapses in the brain, increases [Ca2+]i in cultured rat hippocampal neurons.Amino-acid SequenceCys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH2(Cys1-Cys6)
  • GHS07, Warning