Boc-D-Phe-Pro-OH
Supplier: ALADDIN SCIENTIFIC
B355597-1G
B355597-250MG
ALB355597-1GEA
76.9
USD
ALB355597-1G
ALB355597250MG
Boc-D-Phe-Pro-OH
Proteins and Peptides
sBoc-D-Phe-Pro-OH is used as a reactant in the preparation of Phe-Pro-p-amidinobenzylamine analogs as potent and highly selective thrombin inhibitors.
- GHS07, Warning