Albumin from Bovine Serum
Supplier: ALADDIN SCIENTIFIC
A116563-1G A116563-25G A116563-5G
ALA116563-1GEA 25.9 USD
ALA116563-1G ALA116563-25G ALA116563-5G
Albumin from Bovine Serum
Proteins and Peptides
A general purpose purified albumin, suitable for Westerns, enzyme systems and as a protein supplement.
  • Density: 1.13 g/cm³