Hemoglobin
Supplier: ALADDIN SCIENTIFIC
H195714-1G H195714-25G H195714-5G
ALH195714-1GEA 9.9 USD
ALH195714-1G ALH195714-25G ALH195714-5G
Hemoglobin
Proteins and Peptides
Hemoglobin is the major component of red blood cells, and is responsible for their red color. Its normal concentration in erythrocytes is 34%.Hemoglobin is a globular protein with α and β chains with each 141 and 146 amino acids, respectively. It exists as a tetramer with each monomer having heterocyclic porphyrin ring with iron constituting the heme.Hemoglobin from bovine blood has been used in:• standard curve generation for the quantification intraparenchymal hemorrhage and parenchymal hemorrhage in spinal cord homogenate using Drabkin′s assay, Quadrupole-Ion Mobility-Time-of-Flight mass spectrometery •  the generation of molecularly imprinted polymers (MIPs) to mimic high molecular-weight polyethylene glycol (PEG) in crystallization studiesThe solubility of α-elastin has been applied to construction of elastin-mimetic biomaterials.