H-Gly-Pro-Arg-Pro-OH Acetate Salt
Supplier: ALADDIN SCIENTIFIC
H333592-25MG H333592-5MG H333592-100MG H333592-250MG
ALH333592-25MGEA 276.9 USD
ALH333592-25MG ALH333592-5MG ALH333592100MG ALH333592250MG
H-Gly-Pro-Arg-Pro-OH Acetate Salt
Proteins and Peptides
H-Gly-Pro-Arg-Pro-OH Acetate Salt can bind to the polymerization site (D domain) in fibrinogen (K|i|= 400 mM) and prevent fibrinogen aggregation and fibrin polymerization. It also acts as an anticoagulant. Research shows that there is additional proteolysis of α, β and γ fibrinogen polypeptides during the cleavage of fibrinogen, which may also include the initial removal of fibrinopeptides.