Bombesin acetate
Supplier: ALADDIN SCIENTIFIC
B118773-100MG B118773-1MG B118773-25MG B118773-5MG
ALB118773100MGEA 2662.9 USD
ALB118773100MG ALB118773-1MG ALB118773-25MG ALB118773-5MG
Bombesin acetate
Proteins and Peptides
pGlu-Gln-Arg-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2Bombesin is a tetradeceptide that plays an important role in gastrin release and G protein receptor activation;A neuropeptide with diverse biological activitiesApplication:Gut tetradecapeptide with the ability to stimulate release of various hormones