Angiotensin III, human, mouse (Acetate)
Supplier: ALADDIN SCIENTIFIC
A302486-100MG A302486-25MG A302486-5MG
ALA302486-100MGEA 687.9 USD
ALA302486-100MG ALA302486-25MG ALA302486-5MG
Angiotensin III, human, mouse (Acetate)
Proteins and Peptides
Angiotensin III, human, mouse is a heptapeptide, acts as an endogenous angiotensin type 2 receptor (AT2R) agonist, with IC50s of 0.648 nM and 21.1 nM for AT2R and AT1R, respectively.