Acetyl coenzyme A trilithium salt

Supplier: ALADDIN SCIENTIFIC

A336151-1MG A336151-25MG A336151-5MG
ALA336151-1MGEA 54.9 USD
ALA336151-1MG ALA336151-25MG ALA336151-5MG
Acetyl coenzyme A trilithium salt
Proteins and Peptides

Acetyl Coenzyme A Trilithium Salt Trihydrate is an essential cofactor and carrier of acyl groups in enzymatic acetyl transfer reactions. It is also the precursor for lipid biosynthesis and the neurotransmitter acetylcholine.Carrier of acyl groups for enzymatic reactions involving fatty acid synthesis and oxidation and in biological acetylations.Carrier of acyl groups in enzymatic reactions in fatty acid synthesis and oxidation and in biological acetylations.


  • GHS07, Warning
  • EINECS: 200-790-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR