Bradykinin
Supplier: ALADDIN SCIENTIFIC
B304069-10MG B304069-50MG
ALB304069-10MGEA 79.9 USD
ALB304069-10MG ALB304069-50MG
Bradykinin
Proteins and Peptides
Bradykinin is an active peptide that is generated by the kallikrein-kinin system. It is a inflammatory mediator and also recognized as a neuromediator and regulator of several vascular and renal functions.
  • EINECS: 200-398-8