Beauvericin
Supplier: ALADDIN SCIENTIFIC
B135407-10MG B135407-1MG B135407-25MG B135407-5MG
ALB135407-10MGEA 338.9 USD
ALB135407-10MG ALB135407-1MG ALB135407-25MG ALB135407-5MG
Beauvericin
Proteins and Peptides
Beauvercin is a mycotoxin within the enniatin family produced by Fusarium sp. As an enniatin mycotoxin, Beauvercin has been reported to affect mitochondria through depletion of the transmembrane potential, induction of swelling and in decreasing calcium retention capacity. A screening method for quantification of Beauvercin and other mycotoxins in aqueous environmental samples has been described involving solid-phase extraction, liquid chromatography and mass spectometry.An enniatin family mycotoxin