Guanidine thiocyanate
Supplier: CHEM-IMPEX INTERNATIONAL, INC
Guanidine thiocyanate is a versatile chemical compound widely utilized in molecular biology and biochemistry, particularly for its role in nucleic acid extraction and purification. This compound acts as a chaotropic agent, effectively disrupting hydrogen bonds and facilitating the denaturation of proteins and nucleic acids. Its unique properties make it an essential reagent in the isolation of RNA and DNA from biological samples, ensuring high yields and purity. Researchers appreciate its efficiency in lysing cells and inactivating RNases, which is crucial for maintaining the integrity of nucleic acids during extraction processes.
In addition to its applications in nucleic acid research, guanidine thiocyanate is also employed in various industrial processes, including the synthesis of pharmaceuticals and agrochemicals. Its ability to solubilize proteins and enhance reaction rates makes it a valuable component in formulations requiring protein stabilization. With its broad range of applications and effectiveness in molecular biology, guanidine thiocyanate stands out as a reliable choice for researchers and industry professionals seeking high-quality results in their work.
UN 1759
Packing Group III
Melting Point: 116 - 121 °C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...