Amyloid β Protein Fragment 1-42, TFA
Supplier: ALADDIN SCIENTIFIC
A494432-1MG A494432-500UG
ALA494432-1MGEA 279.9 USD
ALA494432-1MG ALA494432500UG
Amyloid β Protein Fragment 1-42, TFA
Proteins and Peptides
Sequence:Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-AlaBiochemical mechanism:Amyloid protein β Protein segment 1-42 (A β  1-42) It has antioxidant and neuroprotective properties. Amyloid protein β Protein accumulation is associated with Alzheimer's disease (AD) and Down syndrome. A β  1-42 regulates cholesterol transport and acts as a transcription factor. It may also have anti-inflammatory and antimicrobial effects.Application:Amyloid protein is found in the brain of patients with Alzheimer's disease and Down syndrome β- The main segment of the protein.Amyloid protein β Protein fragments 1-42 have been used to:1. A β  Preparation of 1-42 oligomer2. Western blot analysis3. Immunomagnetic Reduction (IMR) Plasma A β 42 Detected interference test4. Study the effect of resveratrol on A β  1-42 induced impairment of spatial learning, memory and synaptic plasticity5. Study A β Role in epithelial cell culture