PRIME-XV FreezIS DMSO-Free excipient
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
DMSO-free, protein-free, animal component-free, excipient chemically defined cryopreservation solution.
DMSO-free, rotein-free, animal component-free, chemically defined cryopreservation solution
Enables cell preservation for short-term at −80 °C and long-term storage in liquid nitrogen to −196 °C
High post-thaw viability and growth
Maintains cell surface marker expression of mesenchymal stem cells (MSCs) and T cells post-thaw
Complete, ready-to-use medium
Designed and manufactured to facilitate transfer from research to clinic. Supported by robust raw material controls and supply chain management
Certificaciones: FDA, Federal, and State registered - cGMP-compliant manufacture; EN ISO 13485:2016 certified; MDSAP certified; Extensive QC testing including functionality, sterility (USP <71>), endotoxin (USP <85>), and mycoplasma (USP <63>); Drug Master Files (DMFs) filed with the FDA – available upon request; Excipient Use Agreement (EUA) will need to be signed for regulatory support.