Anti-ATP citrate lyase (phospho S451 + T447) Rabbit Monoclonal Antibody [clone: EP737Y]

Supplier: Abcam
AB247346-100UG AB247346-1MG
ABCA_AB247346-100UEA 1134 USD
ABCA_AB247346-100U ABCA_AB247346-1MG
Anti-ATP citrate lyase (phospho S451 + T447) Rabbit Monoclonal Antibody [clone: EP737Y]
Antibodies
Rabbit monoclonal [EP737Y] to ATP citrate lyase (phospho T447 + S451) - BSA and Azide free.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ATP citrate lyase
Clonality: Monoclonal
Clone: EP737Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR