Anti-Macrophage Inflammatory Protein 3 alpha Rabbit Monoclonal Antibody [clone: EPR22376-58]

Supplier: Abcam
AB224188-1ML AB224188-100UL
ABCA_AB224188-1MLEA 6329 USD
ABCA_AB224188-1ML ABCA_AB224188-100U
Anti-Macrophage Inflammatory Protein 3 alpha Rabbit Monoclonal Antibody [clone: EPR22376-58]
Antibodies
Rabbit monoclonal [EPR22376-58] to Macrophage Inflammatory Protein 3 alpha.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Macrophage Inflammatory Protein 3 alpha
Clonality: Monoclonal
Clone: EPR22376-58
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR