Anti-CNGA1 Mouse Monoclonal Antibody [clone: PMc 1D1]

Supplier: Abcam
AB255766-100UG AB255766-1MG
ABCA_AB255766-100UEA 1134 USD
ABCA_AB255766-100U ABCA_AB255766-1MG
Anti-CNGA1 Mouse Monoclonal Antibody [clone: PMc 1D1]
Antibodies
Mouse monoclonal [PMc 1D1] to CNGA1 - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CNGA1
Clonality: Monoclonal
Clone: PMc 1D1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR