Anti-Kappa light chain Rabbit Recombinant Multiclonal Antibody [clone: RM1057]

Supplier: Abcam
AB313895-1ML AB313895-20UL AB313895-100UL
ABCA_AB313895-1MLEA 5985 USD
ABCA_AB313895-1ML ABCA_AB313895-20UL ABCA_AB313895-100U
Anti-Kappa light chain Rabbit Recombinant Multiclonal Antibody [clone: RM1057]
Antibodies
Rabbit recombinant multiclonal [RM1057] to Kappa light chain.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Kappa light chain
Clonality: Recombinant Multiclonal
Clone: RM1057
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR