Anti-Gastric Mucin/MUC-6 Rabbit Monoclonal Antibody [clone: EPR20623]

Supplier: Abcam
AB223846-20UL AB223846-100UL AB223846-1ML
ABCA_AB223846-20ULEA 213 USD
ABCA_AB223846-20UL ABCA_AB223846-100U ABCA_AB223846-1ML
Anti-Gastric Mucin/MUC-6 Rabbit Monoclonal Antibody [clone: EPR20623]
Antibodies
Rabbit monoclonal [EPR20623] to Gastric Mucin/MUC-6.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Gastric Mucin/MUC-6
Clonality: Monoclonal
Clone: EPR20623
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR