Anti-Calcium channel L type DHPR alpha 2 subunit/CACNA2D1 Rabbit Monoclonal Antibody [clone: EPR23267-8]

Supplier: Abcam
AB253190-100UL AB253190-1ML
ABCA_AB253190-100UEA 708 USD
ABCA_AB253190-100U ABCA_AB253190-1ML
Anti-Calcium channel L type DHPR alpha 2 subunit/CACNA2D1 Rabbit Monoclonal Antibody [clone: EPR23267-8]
Antibodies
Rabbit monoclonal [EPR23267-8] to Calcium channel L type DHPR alpha 2 subunit/CACNA2D1.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Calcium channel L type DHPR alpha 2 subunit/CACNA2D1
Clonality: Monoclonal
Clone: EPR23267-8
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR