Anti-Myosin light chain 3 Rabbit Monoclonal Antibody [clone: YCA-R22092-208 H4-2L1-2]

Supplier: Abcam
AB288607-1MG AB288607-100UG
ABCA_AB288607-1MGEA 9770 USD
ABCA_AB288607-1MG ABCA_AB288607-100U
Anti-Myosin light chain 3 Rabbit Monoclonal Antibody [clone: YCA-R22092-208 H4-2L1-2]
Antibodies
Rabbit monoclonal [YCA-R22092-208 H4-2L1-2] to Myosin light chain 3 - BSA and Azide free (Capture).

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Myosin light chain 3
Clonality: Monoclonal
Clone: YCA-R22092-208 H4-2L1-2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR