Anti-PD-L2 Mouse Monoclonal Antibody [clone: CAL28]

Supplier: Abcam
AB283344-100UL AB283344-20UL AB283344-1ML
ABCA_AB283344-100UEA 739 USD
ABCA_AB283344-100U ABCA_AB283344-20UL ABCA_AB283344-1ML
Anti-PD-L2 Mouse Monoclonal Antibody [clone: CAL28]
Antibodies
Mouse monoclonal [CAL28] to PD-L2 - Mouse IgG1.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PD-L2
Clonality: Monoclonal
Clone: CAL28
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR