Anti-Pancreatic Polypeptide Rabbit Monoclonal Antibody [clone: EPR23320-150]

Supplier: Abcam
AB281153-100UG AB281153-1MG
ABCA_AB281153-100UEA 990 USD
ABCA_AB281153-100U ABCA_AB281153-1MG
Anti-Pancreatic Polypeptide Rabbit Monoclonal Antibody [clone: EPR23320-150]
Antibodies
Rabbit monoclonal [EPR23320-150] to Pancreatic Polypeptide - BSA and Azide free (Capture).

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Pancreatic Polypeptide
Clonality: Monoclonal
Clone: EPR23320-150
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR