Anti-p21 Rabbit Monoclonal Antibody [clone: EPR3993]

Supplier: Abcam
AB215971-100UG AB215971-1MG
ABCA_AB215971-100UEA 1134 USD
ABCA_AB215971-100U ABCA_AB215971-1MG
Anti-p21 Rabbit Monoclonal Antibody [clone: EPR3993]
Antibodies
Rabbit monoclonal [EPR3993] to p21 - BSA and Azide free.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: p21
Clonality: Monoclonal
Clone: EPR3993
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR