Anti-KLRF1 Rabbit Monoclonal Antibody [clone: EPR23633-150] (PE (Phycpoerythrin))

Supplier: Abcam
AB315145-100UL AB315145-10UL
ABCA_AB315145-100UEA 790 USD
ABCA_AB315145-100U ABCA_AB315145-10UL
Anti-KLRF1 Rabbit Monoclonal Antibody [clone: EPR23633-150] (PE (Phycpoerythrin))
Antibodies
PE Rabbit monoclonal [EPR23633-150] to KLRF1.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: KLRF1
Clonality: Monoclonal
Clone: EPR23633-150
Conjugation: PE (Phycpoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR