Anti-CEACAM6 Rabbit Monoclonal Antibody [clone: 408] PE (Phycoerythrin)

Supplier: Abcam
AB275676-100TEST AB275676-25TEST
ABCA_AB275676-100TEA 696 USD
ABCA_AB275676-100T ABCA_AB275676-25TE
Anti-CEACAM6 Rabbit Monoclonal Antibody [clone: 408] PE (Phycoerythrin)
Antibodies
PE Rabbit monoclonal [408] to CEACAM6.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CEACAM6
Clonality: Monoclonal
Clone: 408
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR