Anti-DDX4 / MVH Rabbit Monoclonal Antibody [clone: EPR24148-58]

Supplier: Abcam
AB270534-20UL AB270534-1ML AB270534-100UL
ABCA_AB270534-20ULEA 295 USD
ABCA_AB270534-20UL ABCA_AB270534-1ML ABCA_AB270534-100U
Anti-DDX4 / MVH Rabbit Monoclonal Antibody [clone: EPR24148-58]
Antibodies
Rabbit monoclonal [EPR24148-58] to DDX4 / MVH.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: DDX4 / MVH
Clonality: Monoclonal
Clone: EPR24148-58
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR