Anti-TARC/CCL17 Rabbit Monoclonal Antibody [clone: YCA-R21766-150 H4L3]

Supplier: Abcam
AB288599-1MG AB288599-100UG
ABCA_AB288599-1MGEA 9770 USD
ABCA_AB288599-1MG ABCA_AB288599-100U
Anti-TARC/CCL17 Rabbit Monoclonal Antibody [clone: YCA-R21766-150 H4L3]
Antibodies
Rabbit monoclonal [YCA-R21766-150 H4L3] to TARC/CCL17 - BSA and Azide free (Capture).

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TARC/CCL17
Clonality: Monoclonal
Clone: YCA-R21766-150 H4L3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR