Anti-MAdCAM1 Rabbit Monoclonal Antibody [clone: EPR27223-58] (Alexa Fluor® 647)

Supplier: Abcam
AB318244-10UL AB318244-100UL
ABCA_AB318244-10ULEA 232 USD
ABCA_AB318244-10UL ABCA_AB318244-100U
Anti-MAdCAM1 Rabbit Monoclonal Antibody [clone: EPR27223-58] (Alexa Fluor® 647)
Antibodies
Alexa Fluor® 647 Rabbit monoclonal [EPR27223-58] to MAdCAM1.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: MAdCAM1
Clonality: Monoclonal
Clone: EPR27223-58
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR