Anti-ERAS Rabbit Monoclonal Antibody [clone: EPR15595]

Supplier: Abcam
AB239011-1MG AB239011-100UG
ABCA_AB239011-1MGEA 10227 USD
ABCA_AB239011-1MG ABCA_AB239011-100U
Anti-ERAS Rabbit Monoclonal Antibody [clone: EPR15595]
Antibodies
Rabbit monoclonal [EPR15595] to ERAS - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ERAS
Clonality: Monoclonal
Clone: EPR15595
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR