Anti-EZHIP Rabbit Monoclonal Antibody [clone: EPR28397-58]

Supplier: Abcam
AB316158-100UG AB316158-1MG
ABCA_AB316158-100UEA 1134 USD
ABCA_AB316158-100U ABCA_AB316158-1MG
Anti-EZHIP Rabbit Monoclonal Antibody [clone: EPR28397-58]
Antibodies
Rabbit monoclonal [EPR28397-58] to EZHIP - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: EZHIP
Clonality: Monoclonal
Clone: EPR28397-58
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR