Anti-PKC zeta+PKC iota Rabbit Monoclonal Antibody [clone: EP1491Y]

Supplier: Abcam
AB247446-1MG AB247446-100UG
ABCA_AB247446-1MGEA 10227 USD
ABCA_AB247446-1MG ABCA_AB247446-100U
Anti-PKC zeta+PKC iota Rabbit Monoclonal Antibody [clone: EP1491Y]
Antibodies
Rabbit monoclonal [EP1491Y] to PKC zeta (phospho T410) + PKC iota (phospho T412) - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PKC zeta+PKC iota
Clonality: Monoclonal
Clone: EP1491Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR