Paragon Scientific Medical Grade Viscosity, 4.0 mPa.s @ 25 °C - contains antibacterial agent
Supplier: LGC Standards
ALK-MGVS40-100 ALK-MGVS40-500
77975-894EA 58 USD
77975-894 77975-895
Paragon Scientific Medical Grade Viscosity, 4.0 mPa.s @ 25 °C - contains antibacterial agent
Standards Viscometry Standards
  • Standard type: Single component