Paragon Scientific Vapour Pressure ASTM D5191: DVPE at 51.6 kPa, Ptot at 57.3 kPa
Supplier: LGC Standards
ALK-CRMU-VP04 ALK-CRMU-VP04K
77975-575EA 636 USD
77975-575 77975-576
Paragon Scientific Vapour Pressure ASTM D5191: DVPE at 51.6 kPa, Ptot at 57.3 kPa
Standards Petroleum Industry Standards
  • Shipping conditions: Room temperature