Paragon Scientific Density 80 °C (Nominal: 0.8246 g/ml @ 80 °C)
Supplier: LGC Standards
ALK-DEN80-02
ALK-DEN80-02-500ML
77975-726EA
97
USD
77975-726
77975-727
Paragon Scientific Density 80 °C (Nominal: 0.8246 g/ml @ 80 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...