Paragon Scientific Density 40 °C (Nominal: 0.7934 g/ml @ 40 °C)
Supplier: LGC Standards
ALK-DEN40-03
ALK-DEN40-03-500ML
77975-706EA
97
USD
77975-706
77975-707
Paragon Scientific Density 40 °C (Nominal: 0.7934 g/ml @ 40 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...