Paragon Scientific Density 15 °C (Nominal: 1.0687 g/ml @ 15 °C)
Supplier: LGC Standards
ALK-DEN15-11
ALK-DEN15-11-500ML
77975-655EA
97
USD
77975-655
77975-656
Paragon Scientific Density 15 °C (Nominal: 1.0687 g/ml @ 15 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...