Paragon Scientific Density 60 °C (Nominal: 0.7924 g/ml @ 60 °C)
Supplier: LGC Standards
ALK-DEN60-01
ALK-DEN60-01-500ML
77975-718EA
97
USD
77975-718
77975-719
Paragon Scientific Density 60 °C (Nominal: 0.7924 g/ml @ 60 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...