Anti-TRIM69 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74600-100UL A74600-50UL
77849-892EA 330 USD
77849-892 77849-893
Anti-TRIM69 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:35 to 1:150

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR