Human Albumin (from Serum)
Supplier: MilliporeSigma
12668-100GM 12668-10GM
80601-606EA 2472.46 USD
80601-606 80601-608
Human Albumin (from Serum)
Proteins and Peptides
Human serum albumin (HSA), a globular, soluble, and unglycosylated monomeric protein, is the most abundant protein in the plasma. It is a multidomain protein consisting of three homologous domains, named I, II, and III. Each of these domains contains subdomains A and B possessing common structural motifs. The HSA gene is present on the human chromosome location 4q13.3.